6-Chloro-4-ethoxypyridin-3-amine 95%

Supplier: Ambeed
Warning

A108343-100MG A108343-250MG A108343-5G
77257-528EA 89.61 USD
77257-528 77257-530 77257-532
6-Chloro-4-ethoxypyridin-3-amine 95%
6-Chloro-4-ethoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
MDL Number: MFCD28666482
CAS Number: 872282-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR