(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine (2R,3R)-2,3-dihydroxysuccinate 97%

Supplier: Ambeed
Warning

A108302-10G A108302-1G A108302-25G
77245-558EA 120.51 USD
77245-558 77245-560 77245-562
(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine (2R,3R)-2,3-dihydroxysuccinate 97%
(S)-2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine (2R,3R)-2,3-dihydroxysuccinate

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₇
MW: 452.88 g/mol
MDL Number: MFCD17019358
CAS Number: 210095-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR