Methyl 5-(aminomethyl)nicotinate hydrochloride 97%

Supplier: Ambeed
Warning

A108186-1G A108186-250MG A108186-5G
77233-316EA 285.31 USD
77233-316 77233-318 77233-320
Methyl 5-(aminomethyl)nicotinate hydrochloride 97%
Methyl 5-(aminomethyl)nicotinate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O₂
MW: 202.63 g/mol
MDL Number: MFCD09838869
CAS Number: 1260791-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR