BAY 58-2667 (free base) 98%

Supplier: Ambeed
Warning

A108170-10MG A108170-1MG A108170-25MG A108170-5MG
77233-308EA 220 USD
77233-308 77233-310 77233-312 77233-314
BAY 58-2667 (free base) 98%
BAY 58-2667 (free base)

Precaución: for Research Use Only

Formula: C₃₆H₃₉NO₅
MDL Number: MFCD18782678
CAS Number: 329773-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR