rel-(6aS,12aS)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-ol 98%

Supplier: Ambeed
Warning

A108101-10MG A108101-1MG A108101-25MG A108101-5MG
77245-544EA 76 USD
77245-544 77245-546 77245-548 77245-550
rel-(6aS,12aS)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-ol 98%
rel-(6aS,12aS)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MDL Number: MFCD00270457
CAS Number: 19908-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR