2-Chloro-5-fluoro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A107986-100MG A107986-10G A107986-1G A107986-250MG A107986-5G A107986-25G
77257-516EA 5 USD
77257-516 77257-518 77257-520 77257-522 77257-524 77614-822
2-Chloro-5-fluoro-4-iodopyridine 98%
2-Chloro-5-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClFIN
MDL Number: MFCD03095306
CAS Number: 884494-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR