2-Chloro-3-(chloromethyl)quinoline 98%

Supplier: Ambeed
Danger

A107945-1G A107945-250MG A107945-5G
77257-510EA 36.05 USD
77257-510 77257-512 77257-514
2-Chloro-3-(chloromethyl)quinoline 98%
2-Chloro-3-(chloromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂N
MW: 212.0753 g/mol
MDL Number: MFCD05865134
CAS Number: 90097-52-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR