(4-Bromophenyl)diphenylphosphine oxide 97%

Supplier: Ambeed
Warning

A107615-10G A107615-1G A107615-25G A107615-5G
77245-504EA 55 USD
77245-504 77245-506 77245-508 77614-814
(4-Bromophenyl)diphenylphosphine oxide 97%
(4-Bromophenyl)diphenylphosphine oxide

Precaución: for Research Use Only

Formula: C₁₈H₁₄BrOP
MDL Number: MFCD00411450
CAS Number: 5525-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR