4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane 97%

Supplier: Ambeed
Warning

Synonyms: Parylene F Dimer, 5,6,11,12,13,14,15,16-Octafluorotricyclo[8.2.2.2(4,7)]hexadeca-4,6,10,12,13,15-hexaene

A107571-1G A107571-25G
77257-500EA 19 USD
77257-500 77257-502
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane 97%
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane

Precaución: for Research Use Only

Formula: C₁₆H₈F₈
MW: 352.23 g/mol
MDL Number: MFCD13195584
CAS Number: 1785-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR