4-Bromo-3,5-dichloropyridine 98%

Supplier: Ambeed
Warning

A107518-100G A107518-10G A107518-1G A107518-25G A107518-5G
77283-618EA 202.91 USD
77283-618 77283-620 77283-622 77283-624 77283-626
4-Bromo-3,5-dichloropyridine 98%
4-Bromo-3,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MDL Number: MFCD06797978
CAS Number: 343781-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR