4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione 97%

Supplier: Ambeed
Warning

Synonyms: 3,4,7,8-Tetrahydroquinoline-2,5(1H,6H)-dione

A107397-1G A107397-250MG A107397-5G
77283-594EA 16 USD
77283-594 77283-596 77283-598
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione 97%
4,6,7,8-Tetrahydro-2,5(1H,3H)-quinolinedione

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD01111799
CAS Number: 5057-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR