2-Bromo-1-chloro-4-trifluoromethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-chloro-4-(trifluoromethoxy)benzene

A107178-100G A107178-10G A107178-25G A107178-5G
77257-492EA 115.36 USD
77257-492 77257-494 77257-496 77257-498
2-Bromo-1-chloro-4-trifluoromethoxybenzene 98%
2-Bromo-1-chloro-4-trifluoromethoxybenzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MW: 275.46 g/mol
MDL Number: MFCD06797751
CAS Number: 468075-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR