1-Phenyl-1H-pyrazole-5-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A107038-100MG
A107038-1G
77233-286EA
29.87
USD
77233-286
77233-288
1-Phenyl-1H-pyrazole-5-carbaldehyde 97%
1-Phenyl-1H-pyrazole-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O MW: 172.1800 g/mol |
MDL Number:
MFCD00103497 CAS Number: 132274-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...