2-(Methoxy(4-phenoxyphenyl)methylene)malononitrile 97%

Supplier: Ambeed
Warning

A106970-1G A106970-250MG A106970-25G
77245-482EA 54.59 USD
77245-482 77245-484 77245-486
2-(Methoxy(4-phenoxyphenyl)methylene)malononitrile 97%
2-(Methoxy(4-phenoxyphenyl)methylene)malononitrile

Precaución: for Research Use Only

Formula: C₁₇H₁₂N₂O₂
MW: 276.29 g/mol
MDL Number: MFCD20140315
CAS Number: 330792-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR