1-Methyl-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A106966-100G A106966-10G A106966-1G A106966-25G A106966-500G A106966-5G
77233-270EA 83 USD
77233-270 77233-272 77233-274 77233-276 77233-278 77233-280
1-Methyl-1H-1,2,4-triazole 98%
1-Methyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₃H₅N₃
MDL Number: MFCD01076192
CAS Number: 6086-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR