Methyl 6-bromo-5-methylpyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A106960-100MG A106960-1G A106960-250MG
77257-480EA 51.5 USD
77257-480 77257-482 77257-484
Methyl 6-bromo-5-methylpyrazine-2-carboxylate 97%
Methyl 6-bromo-5-methylpyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.04 g/mol
MDL Number: MFCD24849554
CAS Number: 1166827-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR