3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A106829-1G A106829-250MG A106829-5G
77257-464EA 243.08 USD
77257-464 77257-466 77257-468
3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid 97%
3-(Methoxycarbonyl)cyclopentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.17 g/mol
MDL Number: MFCD19688558
CAS Number: 84545-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR