(E)-(2R,3R,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-5-hydroxy-2-(hydroxymethyl)-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate 97%

Supplier: Ambeed
Warning

Synonyms: plantamajoside

A106743-10MG A106743-1MG A106743-5MG
77245-470EA 68 USD
77245-470 77245-472 77245-474
(E)-(2R,3R,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-5-hydroxy-2-(hydroxymethyl)-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate 97%
(E)-(2R,3R,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-5-hydroxy-2-(hydroxymethyl)-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₂₉H₃₆O₁₆
MDL Number: MFCD20527298
CAS Number: 104777-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR