Basmisanil 98+%

Supplier: Ambeed
Warning

Synonyms: (1,1-Dioxidothiomorpholino)(6-((3-(4-fluorophenyl)-5-methylisoxazol-4-yl)methoxy)pyridin-3-yl)methanone

A106679-10MG A106679-1MG A106679-25MG A106679-5MG A106679-50MG
77257-452EA 118.45 USD
77257-452 77257-454 77257-456 77257-458 77747-794
Basmisanil 98+%
Basmisanil

Precaución: for Research Use Only

Formula: C₂₁H₂₀FN₃O₅S
MW: 445.46 g/mol
MDL Number: MFCD28902245
CAS Number: 1159600-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR