Ethyl 5-bromoisoxazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A106644-100MG A106644-250MG A106644-1G
77257-448EA 320.33 USD
77257-448 77257-450 77565-866
Ethyl 5-bromoisoxazole-3-carboxylate 97%
Ethyl 5-bromoisoxazole-3-carboxylate

Precaución: For research use only.

Formula: C₆H₆BrNO₃
MW: 220.02 g/mol
MDL Number: MFCD32214869
CAS Number: 1914946-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR