2,6-Dichloro-3-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A106570-100G A106570-10G A106570-1G A106570-25G
77245-458EA 307 USD
77245-458 77245-460 77245-462 77245-464
2,6-Dichloro-3-nitrobenzonitrile 98%
2,6-Dichloro-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂Cl₂N₂O₂
MDL Number: MFCD00051513
CAS Number: 5866-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR