2-Amino-3,5-dichloro-4-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-4-methylpyridin-2-amine

A106558-100G A106558-1G
77245-454EA 265.74 USD
77245-454 77245-456
2-Amino-3,5-dichloro-4-methylpyridine 97%
2-Amino-3,5-dichloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.0300 g/mol
MDL Number: MFCD00129031
CAS Number: 31430-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR