4-Piperidinemethanol 97%

Supplier: Ambeed
Danger

Synonyms: Piperidin-4-ylmethanol

A106543-10G A106543-25G A106543-100G
77283-482EA 9.27 USD
77283-482 77283-484 77592-140
4-Piperidinemethanol 97%
4-Piperidinemethanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00174228
CAS Number: 6457-49-4
UN: 3263
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR