tert-Butyl 5,6-dihydro-1H-pyrazolo[3,4-b]pyridine-7(4H)-carboxylate 97%

Supplier: Ambeed
Warning

A106514-100MG A106514-250MG
77270-586EA 45.32 USD
77270-586 77270-588
tert-Butyl 5,6-dihydro-1H-pyrazolo[3,4-b]pyridine-7(4H)-carboxylate 97%
tert-Butyl 5,6-dihydro-1H-pyrazolo[3,4-b]pyridine-7(4H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MW: 223.27 g/mol
MDL Number: MFCD31539843
CAS Number: 2109805-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR