(S)-N-((3R,4S,5S)-3-Methoxy-1-((S)-2-((1R,2R)-1-methoxy-2-methyl-3-oxo-3-(((S)-2-phenyl-1-(thiazol-2-yl)ethyl)amino)propyl)pyrrolidin-1-yl)-5-methyl-1-oxoheptan-4-yl)-N,3-dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Monomethyl Auristatin D
A106511-10MG
A106511-1MG
A106511-5MG
A106511-50MG
77233-264EA
103
USD
77233-264
77233-266
77233-268
77745-803
(S)-N-((3R,4S,5S)-3-Methoxy-1-((S)-2-((1R,2R)-1-methoxy-2-methyl-3-oxo-3-(((S)-2-phenyl-1-(thiazol-2-yl)ethyl)amino)propyl)pyrrolidin-1-yl)-5-methyl-1-oxoheptan-4-yl)-N,3-dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamide 98%
(S)-N-((3R,4S,5S)-3-Methoxy-1-((S)-2-((1R,2R)-1-methoxy-2-methyl-3-oxo-3-(((S)-2-phenyl-1-(thiazol-2-yl)ethyl)amino)propyl)pyrrolidin-1-yl)-5-methyl-1-oxoheptan-4-yl)-N,3-dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamide
Precaución: for Research Use Only
Formula:
C₄₁H₆₆N₆O₆S |
MDL Number:
MFCD25976743 CAS Number: 203849-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...