Isophthalaldehyde 98%

Supplier: Ambeed
Warning

A106508-100G A106508-10G A106508-25G A106508-500G A106508-5G
77233-254EA 39 USD
77233-254 77233-256 77233-258 77233-260 77233-262
Isophthalaldehyde 98%
Isophthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₆O₂
MDL Number: MFCD00003372
CAS Number: 626-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR