N-Methyl-1-naphthylmethylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Terbinafine Related Compound A, N-Methyl-1-(naphthalen-1-yl)methanamine hydrochloride, 1-(Methylaminomethyl)naphthalene Hydrochloride

A106502-1G A106502-5G A106502-100G
77270-582EA 6.18 USD
77270-582 77270-584 77747-793
N-Methyl-1-naphthylmethylamine hydrochloride 97%
N-Methyl-1-naphthylmethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MW: 207.70 g/mol
MDL Number: MFCD00012555
CAS Number: 65473-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR