6-Bromopicolinic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
A106467-100G
A106467-10G
A106467-500G
A106467-5G
A106467-25G
77270-574EA
125.66
USD
77270-574
77270-576
77270-578
77270-580
77549-046
6-Bromopicolinic acid 98%
6-Bromopicolinic acid
Precaución: for Research Use Only
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol |
MDL Number:
MFCD00093197 CAS Number: 21190-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...