Dimethyl isobutylmalonate 95%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 2-isobutylmalonate

A106376-10G A106376-5G
77257-442EA 32 USD
77257-442 77257-444 77257-446
Dimethyl isobutylmalonate 95%
Dimethyl isobutylmalonate

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
MDL Number: MFCD00015640
CAS Number: 39520-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR