(S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene ≥98%, ee 99%

Supplier: Ambeed
Danger

A106307-250MG A106307-5G
77245-450EA 22 USD
77245-450 77592-132
(S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene ≥98%, ee 99%
(S)-3,3'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₄H₂₀Br₂O₄
MW: 532.22 g/mol
CAS Number: 142010-87-5
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR