4-Bromothiophene-2-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

A106262-100G A106262-10G A106262-25G A106262-500G A106262-5G
77283-454EA 58.71 USD
77283-454 77283-456 77283-458 77283-460 77565-858
4-Bromothiophene-2-carbaldehyde 98%
4-Bromothiophene-2-carbaldehyde

Precaución: For research use only.

Formula: C₅H₃BrOS
MW: 191.05 g/mol
MDL Number: MFCD00005431
CAS Number: 18791-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR