2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-2-((2-fluoro-4-iodophenyl)amino)benzoic acid

A106226-100MG A106226-1G A106226-250MG A106226-5G
77245-438EA 38.11 USD
77245-438 77245-440 77245-442 77245-444
2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid 95%
2-(2-Fluoro-4-iodoanilino)-3,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₇F₃INO₂
MW: 393.1 g/mol
MDL Number: MFCD08690072
CAS Number: 391211-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR