tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(4-chlorophenyl)piperazine-1-carboxylate

A106126-100MG A106126-1G A106126-250MG A106126-5G
77233-236EA 28.84 USD
77233-236 77233-238 77233-240 77233-242
tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate 95%
tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁ClN₂O₂
MW: 296.7924 g/mol
MDL Number: MFCD08275679
CAS Number: 769944-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR