4,5-Dibromophthalic acid 97%
Supplier: Ambeed
|
Warning
|
A106101-100MG
A106101-1G
A106101-250MG
A106101-5G
77283-440EA
46.35
USD
77283-440
77283-442
77283-444
77283-446
4,5-Dibromophthalic acid 97%
4,5-Dibromophthalic acid
Precaución: for Research Use Only
Formula:
C₈H₄Br₂O₄ |
MDL Number:
MFCD00623825 CAS Number: 24063-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...