1-Bromo-4-dodecylbenzene 97%

Supplier: Ambeed
Warning

A106099-1G A106099-5G
77233-232EA 53.56 USD
77233-232 77233-234
1-Bromo-4-dodecylbenzene 97%
1-Bromo-4-dodecylbenzene

Precaución: for Research Use Only

Formula: C₁₈H₂₉Br
MW: 325.33 g/mol
MDL Number: MFCD00191386
CAS Number: 126930-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR