2-Methyl-4-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methoxy-2-methyl-4-nitrobenzene

A106085-100G A106085-10G A106085-1G A106085-25G A106085-5G
77270-536EA 75.19 USD
77270-536 77270-538 77270-540 77270-542 77270-544
2-Methyl-4-nitroanisole 98%
2-Methyl-4-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD07787583
CAS Number: 50741-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR