Z-D-TIC-OH 97%

Supplier: Ambeed
Warning

Synonyms: Z-D-Tic-OH, Z-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A106056-10G A106056-1G A106056-25G A106056-5G
77283-426EA 29.87 USD
77283-426 77283-428 77283-430 77283-432
Z-D-TIC-OH 97%
Z-D-TIC-OH

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄
MW: 311.337 g/mol
MDL Number: MFCD00672346
CAS Number: 146684-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR