3,5-Dichloropyridine-N-oxide 97%

Supplier: Ambeed
Warning

A105883-1G A105883-25G
77270-526EA 20 USD
77270-526 77270-528
3,5-Dichloropyridine-N-oxide 97%
3,5-Dichloropyridine N-oxide

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
MDL Number: MFCD00955620
CAS Number: 15177-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR