FIN56 98+%

Supplier: Ambeed
Danger

Synonyms: N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulphonamide, N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulphonamid

A105877-10MG A105877-1MG A105877-25MG A105877-50MG A105877-5MG
77270-516EA 62 USD
77270-516 77270-518 77270-520 77270-522 77270-524
FIN56 98+%
FIN56

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃O₅S₂
MW: 517.66 g/mol
MDL Number: MFCD30480929
CAS Number: 1083162-61-1
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR