KN-92 hydrochloride 98%

Supplier: Ambeed
Warning

A105838-10MG A105838-1MG A105838-25MG A105838-5MG
77233-220EA 204.01 USD
77233-220 77233-222 77233-224 77233-226
KN-92 hydrochloride 98%
KN-92 hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₆Cl₂N₂O₃S
MDL Number: MFCD25372041
CAS Number: 1431698-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR