GSK-269962 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: GSK26996B, GSK-269962A, GSK269962, N-(3-((2-(4-Amino-1,2,5-oxadiazol-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-6-yl)oxy)phenyl)-4-(2-morpholinoethoxy)benzamide hydrochloride, N-[3-[[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-6-yl]oxy]phenyl]-4-[2-(4-morpholinyl)ethoxy]-benzamide hydrochloride

A105790-10MG A105790-1MG A105790-25MG A105790-50MG A105790-5MG
77257-424EA 166 USD
77257-424 77257-426 77257-428 77257-430 77257-432
GSK-269962 hydrochloride 98%
GSK-269962 hydrochloride

Precaución: for Research Use Only

Formula: C₂₉H₃₁ClN₈O₅
MDL Number: MFCD31536773
CAS Number: 2095432-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR