2,4-Dichloro-5,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

A105756-1G A105756-250MG A105756-5G
77245-420EA 70.04 USD
77245-420 77245-422 77245-424
2,4-Dichloro-5,6-dimethylpyrimidine 97%
2,4-Dichloro-5,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.0300 g/mol
MDL Number: MFCD09910263
CAS Number: 1780-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR