Methyl-4-methoxysalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-4-methoxybenzoate

A105735-25G A105735-5G
77270-510EA 11.33 USD
77270-510 77270-512 77270-514
Methyl-4-methoxysalicylate 98%
Methyl 4-methoxysalicylate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00008424
CAS Number: 5446-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR