6-Methylpyridine-2,4-diol 97%
Supplier: Ambeed
|
Warning
|
A105734-10G
A105734-1G
A105734-25G
A105734-5G
77245-412EA
19
USD
77245-412
77245-414
77245-416
77245-418
6-Methylpyridine-2,4-diol 97%
6-Methylpyridine-2,4-diol
Precaución: for Research Use Only
Formula:
C₆H₇NO₂ |
MDL Number:
MFCD01860849 CAS Number: 158152-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...