2-Thiophenemethanol 97%

Supplier: Ambeed
Warning

A105721-100G A105721-10G A105721-25G A105721-500G A105721-5G A105721-1000G
77233-210EA 55 USD
77233-210 77233-212 77233-214 77233-216 77233-218 77745-801
2-Thiophenemethanol 97%
2-Thiophenemethanol

Precaución: for Research Use Only

Formula: C₅H₆OS
MDL Number: MFCD00005454
CAS Number: 636-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR