2,3,4,5-Tetrafluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A105624-1G A105624-10G
77245-404EA 24 USD
77245-404 77745-800
2,3,4,5-Tetrafluorobenzyl alcohol 98%
2,3,4,5-Tetrafluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₄F₄O
MDL Number: MFCD01631452
CAS Number: 53072-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR