5-Fluoro-1H-indole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A105601-100MG A105601-250MG
77233-206EA 233.81 USD
77233-206 77233-208
5-Fluoro-1H-indole-4-carboxylic acid 97%
5-Fluoro-1H-indole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.14 g/mol
MDL Number: MFCD16037409
CAS Number: 908600-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR