4,5-Dihydroxyanthraquinone-2-carboxylic acid 98+%

Supplier: Ambeed
Warning

Synonyms: Rhein, Rhubarb Yellow, Cassic Acid, 4,5-Dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid

A105483-100MG A105483-1G A105483-250MG A105483-25MG A105483-5G A105483-25G
77270-486EA 6.18 USD
77270-486 77270-488 77270-490 77270-492 77270-494 77747-786
4,5-Dihydroxyanthraquinone-2-carboxylic acid 98+%
4,5-Dihydroxyanthraquinone-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₈O₆
MW: 284.22 g/mol
MDL Number: MFCD00009618
CAS Number: 478-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR