3-Bromo-o-anisaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methoxybenzaldehyde

A105276-1G A105276-250MG A105276-25G A105276-5G
77270-464EA 17 USD
77270-464 77270-466 77270-468 77270-470
3-Bromo-o-anisaldehyde 95%
3-Bromo-o-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.0400 g/mol
MDL Number: MFCD09834811
CAS Number: 88275-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR