3,5-Difluorosalicylaldehyde 98%

Supplier: Ambeed
Warning

A105218-10G A105218-250MG A105218-5G A105218-1G A105218-25G
77270-458EA 223.51 USD
77270-458 77270-460 77270-462 77570-316 77570-318
3,5-Difluorosalicylaldehyde 98%
3,5-Difluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
MDL Number: MFCD04039224
CAS Number: 63954-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR